Merck
  • Lysophosphatidic acid expression in theca cells depends on the type of bovine ovarian follicle.
Lysophosphatidic acid expression in theca cells depends on the type of bovine ovarian follicle.
Reproduction in domestic animals = Zuchthygiene (2016-09-21)
E Sinderewicz, K Grycmacher, D Boruszewska, I Kowalczyk-Zięba, I Woclawek-Potocka
ABSTRACT
Lysophosphatidic acid (LPA) exerts various actions on the mammalian reproductive system. In cows, LPA stimulates the synthesis and secretion of luteotropic factors in the ovary, which affects the growth and development of ovarian follicles. The role of LPA in granulosa cells, oocyte and oocyte-cumulus complex (COC) has previously been investigated; but its role in the theca layer, which is an important structural and functional component of the ovarian follicle, is still unclear. The goal of this study was to investigate the expression of LPA in theca cells originating from different bovine ovarian follicle types. Theca cells were separated from healthy, transitional and atretic ovarian follicles, based on intrafollicular estradiol: progesterone ratios. LPA concentration in the follicular fluid (FF) in different follicle types was measured, and expression of the enzymes responsible for LPA synthesis (autotaxin [AX], phospholipase A2 [PLA2]) and receptors for LPA (LPAR1-4) were determined. The obtained results confirmed the follicle-type dependent presence of LPA in the FF of the bovine ovarian follicles. The highest concentration of LPA was detected in follicles classified as healthy and dominant. LPAR1-4, PLA2 and AX expression in theca cells in all of the types of follicles examined were detected at mRNA and protein level. These results suggest that theca cells can be a source of LPA synthesis other than granulosa cells and COCs, as well as the target for its action in the bovine ovarian follicle, with PLA2 and LPAR4 playing major roles in LPA synthesis and action.