- Double labeling and simultaneous detection of B- and T cells using fluorescent nano-crystal (q-dots) in paraffin-embedded tissues.
Double labeling and simultaneous detection of B- and T cells using fluorescent nano-crystal (q-dots) in paraffin-embedded tissues.
A double immunohistochemical technique for the simultaneous detection of T- and B cells in paraffin-embedded mice tissues have been developed. This procedure is based on using fluorescent nano-crystals (q-dots). The benefit of using q-dots evolves from their unique fluorescence characteristics advantages: such as broad excitation spectrum, narrow emission band and high photo-bleaching threshold compare to organic fluorophores. T cells antigens (CD3) were stained using antibody-coated q-dots with max emission at 655 nm (GalphaRb-QD655). B cells antigens (CD45R/B220) were stained using streptavidin-coated q-dots with max emission at 585 nm (SA-QD585). The simultaneous detection of T- and B cells was demonstrated in paraffin-embedded lymph node using standard fluorescence microscope.