Merck
  • An epididymis-specific beta-defensin is important for the initiation of sperm maturation.
An epididymis-specific beta-defensin is important for the initiation of sperm maturation.
Nature cell biology (2004-05-04)
Chen Xi Zhou, Yong-Lian Zhang, Liqing Xiao, Min Zheng, Ka Man Leung, Man Yee Chan, Pui Shan Lo, Lai Ling Tsang, Hau Yan Wong, Lok Sze Ho, Yiu Wa Chung, Hsiao Chang Chan
ABSTRACT
Although the role of the epididymis, a male accessory sex organ, in sperm maturation has been established for nearly four decades, the maturation process itself has not been linked to a specific molecule of epididymal origin. Here we show that Bin1b, a rat epididymis-specific beta-defensin with antimicrobial activity, can bind to the sperm head in different regions of the epididymis with varied binding patterns. In addition, Bin1b-expressing cells, either of epididymal origin or from a Bin1b-transfected cell line, can induce progressive sperm motility in immotile immature sperm. This induction of motility is mediated by the Bin1b-induced uptake of Ca(2+), a mechanism that has a less prominent role in maintaining motility in mature sperm. In vivo antisense experiments show that suppressed expression of Bin1b results in reduced binding of Bin1b to caput sperm and in considerable attenuation of sperm motility and progressive movement. Thus, beta-defensin is important for the acquisition of sperm motility and the initiation of sperm maturation.