Merck
  • Evaluation of immunosuppressive function of regulatory T cells using a novel in vitro cytotoxicity assay.
Evaluation of immunosuppressive function of regulatory T cells using a novel in vitro cytotoxicity assay.
Cell & bioscience (2014-01-01)
Linyi Zhang, Jean N Manirarora, Cheng-Hong Wei
ABSTRACT
Naturally occurring regulatory T cells (Tregs) play a pivotal role in the maintenance of self-tolerance due to their intrinsic immunosuppressive activity. Currently, a number of human clinical trials are being conducted to investigate the roles of Tregs in treating various immune-mediated disorders. Traditionally, the suppressive activity of Tregs is measured using either a thymidine incorporation assay, which is a radioactive assay; or CFSE based flow cytometry assay, which requires a relatively large number of cells. Consequently, there is an increasing need to develop novel alternative bioassays that can characterize various aspects of the immunosuppressive function of Tregs in vitro. In this study, using murine clonal CD8(+) T cells specific for an islet antigen as responder T cells, we first established a novel, sensitive and quantitative in vitro luminescence based cell viability assay to measure cytotoxicity. Then we used this assay to measure if Tregs could inhibit the cytotoxicity of CD8 effector T cells. This assay does not involve the use of radioisotopes and only needs relatively low number of Tregs. Since normally Tregs only constitute 5-10% of peripheral CD4(+) T cells, this advantage is noteworthy compared with other methods. With the assay we developed, we demonstrated that regulatory T cells (Tregs) could inhibit the antigen-specific killing of an adherent target cell monolayer by the CD8(+) cytotoxic T cells. We observed more inhibition when Tregs and CD8 killer T cells were incubated during the in vitro activation (stimulation) stage of the cytotoxic T lymphocytes (CTL) than when they were added later at the start of the effector phase. Interestingly, Tregs from B6 mice demonstrated higher suppression of CD8(+) T cell killing than Tregs from NOD mice. Moreover, IL-2/anti-IL-2 mAb complexes induced expansion of Tregs in vivo, as well as enhancing the Treg's suppressive activity per cell. Therefore, this novel non-radioactive, luminescence based cytotoxicity assay mediated by clonal islet antigen-specific CD8 T cells can be used to measure, characterize, and quantitate the immunosuppressive activity of natural Tregs, representing a useful approach to characterize the functions of Tregs in the setting of autoimmune diseases and to elucidate the mechanisms for Treg cell-mediated immunoregulation.
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, ≥95.0% (HPLC)