- The development of endometrial hyperplasia in aged PD-1-deficient female mice.
The development of endometrial hyperplasia in aged PD-1-deficient female mice.
Programmed death-1 (PD-1, Pdcd1)-deficient mice develop different types of autoimmune diseases depending on the mouse strain but its role in uterus development has not been reported. In this study, the expression of PD-1 and its ligands, PD-L1 and PD-L2, in uterine tissues from aged WT mice in a 129svEv-Brd background was analyzed by immunohistochemistry and the uterine morphology between WT and PD-1-/- mice was compared by hematoxylin and eosin staining. The aged PD-1-/- female mice in a 129svEv-Brd rather than Balb/c background develop endometrial hyperplasia. H&E staining showed an increase in the number of glands, neovascularization and an extremely large luminal cavity in aged PD-1-/- uteri. Immunohistochemical assay showed that the expression of PD-1 was observed in glandular/luminal epithelium and cells infiltrating the stroma. Fluorescent double staining demonstrated that PD-1 expresses on CD68+ macrophages, CD3+ T cells, CD16+ monocytes, CD56+ NK cells and CK-18+ epithelial cells, respectively. Additionally, PD-1 co-expresses with vascular endothelial growth factor (VEGF), and PD-1 deficiency resulted in an accumulation of glandular/luminal epithelium derived VEGF, which accelerates the expression of the proliferation-associated protein, proliferating cell nuclear antigen (PCNA), and thus potentially lead to epithelial proliferation in aged PD-1-/- uteri. These findings showed that PD-1 deficiency augments luminal epithelial cell proliferation probably through induced VEGF secretion, suggesting PD-1 plays an important role in controlling the growth and differentiation of the uterine epithelium. The virtual slide(s) for this article can be found here: http://www.diagnosticpathology.diagnomx.eu/vs/5809067461223905.