Merck
  • Potential regulatory role of calsequestrin in platelet Ca(2+) homeostasis and its association with platelet hyperactivity in diabetes mellitus.
Potential regulatory role of calsequestrin in platelet Ca(2+) homeostasis and its association with platelet hyperactivity in diabetes mellitus.
Journal of thrombosis and haemostasis : JTH (2011-11-09)
Z Zhu, H Zhou, X Yu, L Chen, H Zhang, S Ren, Y Wu, Dali Luo
ABSTRACT
Altered Ca(2+) homeostasis contributes significantly to platelet hyperactivity in diabetes mellitus. Calsequestrin (CSQ), as a Ca(2+) buffer protein in the sarcoplasmic reticulum, also regulates the Ca(2+) release process in muscles. We hypothesized that CSQ may be expressed in platelets, but is altered and involved in diabetic platelet Ca(2+) abnormalities and hyperaggregability. CSQ expression in platelets from streptozotocin-induced type 1 diabetes rats, type 2 diabetes volunteers and Goto-Kakizaki rats were analyzed by western blotting and RT-qPCR. Platelet Ca(2+) and aggregation were evaluated with Fura2 and an aggregometer, respectively. Platelets from diabetic patients and rats exhibited increased resting Ca(2+) levels, and hyperactive Ca(2+) and aggregation responses to agonists. This enhanced basal Ca(2+) was largely dependent on intracellular Ca(2+) and insensitive to inositol 1,4,5-trisphosphate receptor (IP(3)R) antagonism. Additionally, the expression of the skeletal CSQ isotype (CSQ-1) was detected in both rat and human platelets, but its levels were significantly lowered in diabetic platelets as compared with normal platelets. Impairment of CSQ by trifluoperazine caused concentration-dependent Ca(2+) release in normal platelets and HEK293 cells. Knocking down CSQ-1 in HEK293 cells resulted in increased leakage of Ca(2+), which was also insensitive to IP(3)R inhibition, and exaggerated Ca(2+) release following carbachol treatment. Downregulation of CSQ-1 in diabetic platelets and impairment of CSQ-1 in normal cells leads to disturbed Ca(2+) release, demonstrating a potential role for CSQ-1 in the regulation of the platelet Ca(2+) release process and a possible causal contribution to diabetic platelet hyperactivity.