Merck
Chemopreventive effects of sandalwood oil on skin papillomas in mice.
European journal of cancer prevention : the official journal of the European Cancer Prevention Organisation (ECP) (1997-08-01)
C Dwivedi, A Abu-Ghazaleh
ABSTRACT
The essential oil, emulsion or paste of sandalwood (Santalum album L) has been used in India as an ayurvedic medicinal agent for the treatment of inflammatory and eruptive skin diseases. In this investigation, the chemopreventive effects of sandalwood oil (5% in acetone, w/v) on 7,12-dimethylbenz(a)anthracene-(DMBA)-initiated and 12-O-tetradecanoyl phorbol-13-acetate(TPA)-promoted skin papillomas, and TPA-induced ornithine decarboxylase (ODC) activity in CD1 mice were studied. Sandalwood oil treatment significantly decreased papilloma incidence by 67%, multiplicity by 96%, and TPA-induced ODC activity by 70%. This oil could be an effective chemopreventive agent against skin cancer.