- A fatal case due to cough syrup abuse.
A fatal case due to cough syrup abuse.
Soudni lekarstvi (2012-11-06)
H Kinoshita, N Tanaka, M Jamal, M Kumihashi, R Okuzono, K Ameno
PMID23121038
ABSTRACT
We describe here a fatal abused case of cough syrup, containing chlorpheniramine and dihydrocodeine. Postmortem blood concentration of chlorpheniramine was above fatal levels, but dihydrocodeine concentration was within a therapeutic ranges, and those drug levels in blood were discussed from the viewpoint of forensic pharmacokinetics. We concluded that the cause death was due to the chlorpheniramine poisoning. cough syrup abuse - chlorpheniramine - dihydrocodeine.
Product Description
Supelco
Dihydrocodeine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®