Merck
  • Development and validation of a high performance liquid chromatographic method for the determination of oxcarbazepine and its main metabolites in human plasma and cerebrospinal fluid and its application to pharmacokinetic study.
Development and validation of a high performance liquid chromatographic method for the determination of oxcarbazepine and its main metabolites in human plasma and cerebrospinal fluid and its application to pharmacokinetic study.
Journal of pharmaceutical and biomedical analysis (2006-09-09)
Vasilios Kimiskidis, Marios Spanakis, Ioannis Niopas, Dimitrios Kazis, Chrysi Gabrieli, Feras Imad Kanaze, Daniil Divanoglou
ABSTRACT
An isocratic reversed-phase HPLC-UV procedure for the determination of oxcarbazepine and its main metabolites 10-hydroxy-10,11-dihydrocarbamazepine and 10,11-dihydroxy-trans-10,11-dihydrocarbamazepine in human plasma and cerebrospinal fluid has been developed and validated. After addition of bromazepam as internal standard, the analytes were isolated from plasma and cerebrospinal fluid by liquid-liquid extraction. Separation was achieved on a X-TERRA C18 column using a mobile phase composed of 20 mM KH(2)PO(4), acetonitrile, and n-octylamine (76:24:0.05, v/v/v) at 40 degrees C and detected at 237 nm. The described assay was validated in terms of linearity, accuracy, precision, recovery and lower limit of quantification according to the FDA validation guidelines. Calibration curves were linear with a coefficient of variation (r) greater than 0.998. Accuracy ranged from 92.3% to 106.0% and precision was between 2.3% and 8.2%. The method has been applied to plasma and cerebrospinal fluid samples obtained from patients treated with oxcarbazepine, both in monotherapy and adjunctive therapy.
Sigma-Aldrich
Octylamine, ≥99.5%
Sigma-Aldrich
Octylamine, 99%