Merck
  • Classical and alternative complement activation on photoreceptor outer segments drives monocyte-dependent retinal atrophy.
Classical and alternative complement activation on photoreceptor outer segments drives monocyte-dependent retinal atrophy.
Scientific reports (2018-05-11)
Kenneth J Katschke, Hongkang Xi, Christian Cox, Tom Truong, Yann Malato, Wyne P Lee, Brent McKenzie, Rommel Arceo, Jianhua Tao, Linda Rangell, Mike Reichelt, Lauri Diehl, Justin Elstrott, Robby M Weimer, Menno van Lookeren Campagne
ABSTRACT
Geographic atrophy (GA), the advanced form of dry age-related macular degeneration (AMD), is characterized by progressive loss of retinal pigment epithelium cells and photoreceptors in the setting of characteristic extracellular deposits and remains a serious unmet medical need. While genetic predisposition to AMD is dominated by polymorphisms in complement genes, it remains unclear how complement activation contributes to retinal atrophy. Here we demonstrate that complement is activated on photoreceptor outer segments (POS) in the retina peripheral to atrophic lesions associated with GA. When exposed to human serum following outer blood-retinal barrier breakdown, POS act as potent activators of the classical and alternative complement pathway. In mouse models of retinal degeneration, classical and alternative pathway complement activation on photoreceptors contributed to the loss of photoreceptor function. This was dependent on C5a-mediated recruitment of peripheral blood monocytes but independent of resident microglia. Genetic or pharmacologic inhibition of both classical and alternative complement C3 and C5 convertases was required to reduce progressive degeneration of photoreceptor rods and cones. Our study implicates systemic classical and alternative complement proteins and peripheral blood monocytes as critical effectors of localized retinal degeneration with potential relevance for the contribution of complement activation to GA.
Sigma-Aldrich
Anti-Rhodopsin Antibody, CT, last 9 amino acids, clone Rho 1D4, clone Rho 1D4, Chemicon®, from mouse