Merck
  • Apico-basal elongation requires a drebrin-E-EB3 complex in columnar human epithelial cells.
Apico-basal elongation requires a drebrin-E-EB3 complex in columnar human epithelial cells.
Journal of cell science (2012-01-26)
Elsa Bazellières, Dominique Massey-Harroche, Magali Barthélémy-Requin, Fabrice Richard, Jean-Pierre Arsanto, André Le Bivic
ABSTRACT
Although columnar epithelial cells are known to acquire an elongated shape, the mechanisms involved in this morphological feature have not yet been completely elucidated. Using columnar human intestinal Caco2 cells, it was established here that the levels of drebrin E, an actin-binding protein, increase in the terminal web both in vitro and in vivo during the formation of the apical domain. Drebrin E depletion was found to impair cell compaction and elongation processes in the monolayer without affecting cell polarity or the formation of tight junctions. Decreasing the drebrin E levels disrupted the normal subapical F-actin-myosin-IIB-βII-spectrin network and the apical accumulation of EB3, a microtubule-plus-end-binding protein. Decreasing the EB3 levels resulted in a similar elongation phenotype to that resulting from depletion of drebrin E, without affecting cell compaction processes or the pattern of distribution of F-actin-myosin-IIB. In addition, EB3, myosin IIB and βII spectrin were found to form a drebrin-E-dependent complex. Taken together, these data suggest that this complex connects the F-actin and microtubule networks apically during epithelial cell morphogenesis, while drebrin E also contributes to stabilizing the actin-based terminal web.
Product Description
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)