Merck
  • LEDGF binds to heat shock and stress-related element to activate the expression of stress-related genes.
LEDGF binds to heat shock and stress-related element to activate the expression of stress-related genes.
Biochemical and biophysical research communications (2001-05-15)
D P Singh, N Fatma, A Kimura, L T Chylack, T Shinohara
ABSTRACT
We have investigated the mechanism by which LEDGF protects cells against environmental stress. Our earlier report showed that a low level of LEDGF was present in the nucleus of most cell types and significant elevation of LEDGF level was induced by heat and oxidative stress. The cells overexpressing LEDGF-activated expression of heat shock proteins and enhanced survival of many cell types. Here we show that LEDGF binds to heat shock element (HSE) and stress-related regulatory element (STRE) to activate the expression of stress-related genes (Hsp27 and alphaB-crystallin). Apparently, HSE and STRE are present in promoters of many stress-related genes. Elevation of many stress-related proteins (STRPs) induced by LEDGF may protect cells against environmental stress. In yeast, it has been demonstrated that a single stress can activate the expression of multiple STRPs. This is known as "cross-protection," and now similar mechanism has been found in mammalian cells and LEDGF plays a vital role in it.