Indium tin oxide coated glass slide, square
Synonym(s):
ITO, ITO coated slide, square, ITO coated slide, rectangular
Linear Formula:
In2O3 · (SnO2)x
CAS Number:
Products
Product Category
Brand
Length (mm)
Quality Segment
Showing 1-6 of 6
Compare
Product Number
Description
Pricing