![[Ile7]-Angiotensin III acetate salt hydrate ≥95% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/178/636/8f34b268-6027-405e-8e2b-d391e5de3da0/640/8f34b268-6027-405e-8e2b-d391e5de3da0.png)
[Ile7]-Angiotensin III acetate salt hydrate
![[Ile7]-Angiotensin III acetate salt hydrate ≥95% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/178/636/8f34b268-6027-405e-8e2b-d391e5de3da0/640/8f34b268-6027-405e-8e2b-d391e5de3da0.png)
Synonym(s):
Arg-Val-Tyr-Ile-His-Pro-Ile
Empirical Formula (Hill Notation):
C43H68N12O9 · xC2H4O2 · yH2O
Molecular Weight:
897.08 (anhydrous free base basis)
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing