25-Hydroxycholecalciferol
Synonym(s):
25-Hydroxyvitamin D3, Vitamin D3, 25-Hydroxy-
Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing