Skip to Content
Merck
BIS-TRIS anhydrous, free-flowing, Redi-Dri™, ≥98.0%

BIS-TRIS

BIS-TRIS anhydrous, free-flowing, Redi-Dri™, ≥98.0%
Synonym(s):
2,2-Bis(hydroxymethyl)-2,2′,2″-nitrilotriethanol, 2-Bis(2-hydroxyethyl)amino-2-(hydroxymethyl)-1,3-propanediol, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane, 2,2-Bis(hydroxyethyl)-(iminotris)- (hydroxymethyl)-methane
Empirical Formula (Hill Notation):
C8H19NO5
CAS Number:
Molecular Weight:
209.24
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Research Area
Quality Segment
Application
Special Grade
Technique