Skip to Content
Merck
Fructose European Pharmacopoeia (EP) Reference Standard

D-(−)-Fructose

Fructose European Pharmacopoeia (EP) Reference Standard
Synonym(s):
D-Levulose, Fruit sugar, D-(−)-Fructose, D(-)-Fructose, D(-)Fructose
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Format
Analyte Suitability
Quality Segment
Application
Markush Class
Markush Group
Microorganism Suitability
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique