Skip to Content
Merck
Cellulose DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)

Cellulose

Cellulose DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Synonym(s):
Cellulose powder, Cotton linters, Cellulose, Cellulose microcrystalline
Linear Formula:
(C6H10O5)n
CAS Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Manufacturer
pH Value
Physical Form
Separation Technique
Length (m)
Length (mm)
Quality Segment
Particle Size (µm)
Polymer Composition
Polymer Type
Special Grade
Technique
Showing 1-20 of 29
Page 1 of 2
Page 1 of 2