Skip to Content
Merck
Trypsin Sequencing Grade from bovine pancreas

Trypsin from porcine pancreas

Trypsin Sequencing Grade from bovine pancreas
Synonym(s):
Trypsin bovine, Trypsin porcine, Trypsin, Bovine Pancreas, Trypsin, Iodination Grade, Human Pancreas, Trypsin
Empirical Formula (Hill Notation):
C1012H1585N279O324S14
CAS Number:
Molecular Weight:
23293.00

Products

Product Category
Brand
Biological Source
Gene Alias
Manufacturer
Physical Form
Product Line
Recombinant Host
Tissue
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-20 of 34
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2