Skip to Content
Merck
Potassium bromide FT-IR grade, ≥99% trace metals basis

Potassium bromide

Potassium bromide FT-IR grade, ≥99% trace metals basis
Synonym(s):
Potassium bromide
Linear Formula:
KBr
CAS Number:
Molecular Weight:
119.00

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Special Grade