Merck
  • Structural basis for specific cleavage of Lys6-linked polyubiquitin chains by USP30.
Structural basis for specific cleavage of Lys6-linked polyubiquitin chains by USP30.
Nature structural & molecular biology (2017-09-26)
Yusuke Sato, Kei Okatsu, Yasushi Saeki, Koji Yamano, Noriyuki Matsuda, Ai Kaiho, Atsushi Yamagata, Sakurako Goto-Ito, Minoru Ishikawa, Yuichi Hashimoto, Keiji Tanaka, Shuya Fukai
ABSTRACT
Parkin ubiquitin (Ub) ligase (also known as PARK2) ubiquitinates damaged mitochondria for their clearance and quality control. USP30 deubiquitinase opposes parkin-mediated Ub-chain formation on mitochondria by preferentially cleaving Lys6-linked Ub chains. Here, we report the crystal structure of zebrafish USP30 in complex with a Lys6-linked diubiquitin (diUb or Ub