Merck
  • Convergent ERK1/2, p38 and JNK mitogen activated protein kinases (MAPKs) signalling mediate catecholoestradiol-induced proliferation of ovine uterine artery endothelial cells.
Convergent ERK1/2, p38 and JNK mitogen activated protein kinases (MAPKs) signalling mediate catecholoestradiol-induced proliferation of ovine uterine artery endothelial cells.
The Journal of physiology (2017-04-25)
Rosalina Villalon Landeros, Sheikh O Jobe, Gabrielle Aranda-Pino, Gladys E Lopez, Jing Zheng, Ronald R Magness
ABSTRACT
The catechol metabolites of 17β-oestradiol (E Previously we demonstrated that the biologically active metabolites of 17β-oestradiol, 2-hydroxyoestradiol (2-OHE
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)
Sigma-Aldrich
SP600125, ≥98% (HPLC)