Merck
  • Deneddylase 1 regulates deneddylase activity of the Cop9 signalosome in Drosophila melanogaster.
Deneddylase 1 regulates deneddylase activity of the Cop9 signalosome in Drosophila melanogaster.
Insect science (2015-09-04)
Kiyoung Kim, Jeongsook Yoon, Jeongbin Yim, Hyung-Jun Kim
ABSTRACT
NEDD8 conjugation of Cullin has an important role in ubiquitin-mediated protein degradation. The COP9 signalosome, of which CSN5 is the major catalytic subunit, is a major Cullin deneddylase. Another deneddylase, Deneddylase 1, has also been shown to process the Nedd8 precursor. In Drosophila, the DEN1 mutants do not have increased levels of Cullin neddylation, but instead show a significant decrease in neddylated Cullin. This characteristic decrease in neddylated Cullins in the DEN1