Merck
  • Understanding functional miRNA-target interactions in vivo by site-specific genome engineering.
Understanding functional miRNA-target interactions in vivo by site-specific genome engineering.
Nature communications (2014-08-20)
Andrew R Bassett, Ghows Azzam, Lucy Wheatley, Charlotte Tibbit, Timothy Rajakumar, Simon McGowan, Nathan Stanger, Philip Andrew Ewels, Stephen Taylor, Chris P Ponting, Ji-Long Liu, Tatjana Sauka-Spengler, Tudor A Fulga
ABSTRACT
MicroRNA (miRNA) target recognition is largely dictated by short 'seed' sequences, and single miRNAs therefore have the potential to regulate a large number of genes. Understanding the contribution of specific miRNA-target interactions to the regulation of biological processes in vivo remains challenging. Here we use transcription activator-like effector nuclease (TALEN) and clustered regularly interspaced short palindromic repeat (CRISPR)/Cas9 technologies to interrogate the functional relevance of predicted miRNA response elements (MREs) to post-transcriptional silencing in zebrafish and Drosophila. We also demonstrate an effective strategy that uses CRISPR-mediated homology-directed repair with short oligonucleotide donors for the assessment of MRE activity in human cells. These methods facilitate analysis of the direct phenotypic consequences resulting from blocking specific miRNA-MRE interactions at any point during development.
Product Description
Roche
NBT, 4-Nitro blue tetrazolium chloride, solution
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep