Merck
  • Comparing transplant glomerulopathy in the absence of C4d deposition and donor-specific antibodies to chronic antibody-mediated rejection.
Comparing transplant glomerulopathy in the absence of C4d deposition and donor-specific antibodies to chronic antibody-mediated rejection.
Clinical transplantation (2014-08-12)
Irina B Torres, Maite Salcedo, Francesc Moreso, Joana Sellarés, Eva Castellá, M Antonieta Azancot, Manel Perelló, Carme Cantarell, Daniel Serón
ABSTRACT
Transplant glomerulopathy (TG) is the characteristic lesion of chronic antibody-mediated rejection (AMR). However, in some patients presents with no circulating HLA antibodies or C4d positivity. Patients with TG accomplishing criteria for chronic AMR were compared to patients with isolated TG. We reviewed late (>6 months) graft biopsies performed between 2007 and 2010 (n = 75). Biopsies with C4d-negative TG and no circulating donor-specific antibody were called isolated TG (n = 12), and chronic AMR was defined according to Banff consensus (n = 17). HLA antibodies were evaluated by Luminex technology. Immunohistochemistry was performed to quantify graft infiltrating cells. Patients with isolated TG were older (52 ± 14 vs. 35 ± 14; p = 0.0048), received grafts from older donors (54 ± 16 vs. 41 ± 18; p = 0.0554), and displayed a lower inflammation in the glomerular (g-score: 0.5 ± 0.5 vs. 1.0 ± 0.9; p = 0.0865; CD3 positive cells/glomeruli: 1.5 ± 2.9 vs. 4.4 ± 4.1; p = 0.0147), interstitial (i-score: 1.2 ± 0.9 vs. 1.9 ± 1.0; p = 0.0685; CD45 positive cells/hpf: 18 ± 11 vs. 57 ± 68; p = 0.0132), and peritubular capillary (ptc-score 0.2 ± 0.6 vs. 1.1 ± 0.9; p = 0.0089; CD45 positive cells/hpf: 3.7 ± 3.1 vs. 10.1 ± 7.4; p = 0.0290) compartments. Fifteen grafts were lost and graft survival was significantly lower in patients with chronic AMR (p = 0.0122). Isolated TG is associated with less severe allograft inflammation and with a better outcome than chronic AMR.