Merck
  • Fas, FasL, and cleaved caspases 8 and 3 in glioblastomas: a tissue microarray-based study.
Fas, FasL, and cleaved caspases 8 and 3 in glioblastomas: a tissue microarray-based study.
Pathology, research and practice (2014-02-25)
Fabiano P Saggioro, Luciano Neder, João Norberto Stávale, Aline Nazareth P Paixão-Becker, Suzana M F Malheiros, Fernando A Soares, José Eymard H Pittella, Caio César M S Matias, Benedicto O Colli, Carlos Gilberto Carlotti, Marcello Franco
ABSTRACT
This investigation analyzed the immunoexpression of FasL, Fas, cleaved caspase-8, and cleaved caspase-3 in glioblastomas. Formalin-fixed and paraffin-embedded glioblastoma tissues and control brain tissues from 97 patients were analyzed by tissue microarrays and immunohistochemistry. Patients with glioblastomas that were negative or weakly stained (<50% of cells positive) for cleaved caspase-8 had worse cancer-specific overall survival (median=8.5 months) than did patients with tumors that highly expressed cleaved caspase-8 (median=11.7 months; P=0.0325), independent of clinical variables. There was no association of other markers with survival, treatment, sex, age, tumor size, and primary site. Among the tumors, there were reasonable to good positive correlations between the expression of FasL and Fas (r=0.47) and between Fas and cleaved caspase-8 (r=0.41), and there were poor positive correlations between Fas and cleaved caspase-3 (r=0.26), FasL and cleaved caspase-8 (r=0.22), and cleaved caspase-8 and -3 (r=0.31). Our results suggest that Fas-Fas-ligand signal transduction could be inhibited, especially at the stage of caspase-8 activation, thereby establishing a major mechanism for evasion of apoptosis by these tumors. The absence or low expression of cleaved caspase-8 in the tumors was a negative prognostic indicator for patient survival.