Merck
  • Electro-optical Synergy Technique: A New and Effective Nonablative Approach to Skin Aging.
Electro-optical Synergy Technique: A New and Effective Nonablative Approach to Skin Aging.
The Journal of clinical and aesthetic dermatology (2011-01-05)
Moetaz El-Domyati, Tarek S El-Ammawi, Walid Medhat, Osama Moawad, My G Mahoney, Jouni Uitto
ABSTRACT
Electro-optical synergy technology is one of the most recently described methods for nonablative skin rejuvenation. The aim of this study is to evaluate the effects of electro-optical synergy on connective tissue composition by histological and immunohistochemical techniques coupled with computerized morphometric analysis. A prospective clinical study. Six volunteers with Fitzpatrick skin types 3 to 4 and Glogau class I to II wrinkles were subjected to three months (6 sessions at 2-week intervals) of electro-optical synergy treatment. Standard photographs and skin biopsies were obtained at baseline as well as three and six months after the start of treatment. The authors performed quantitative evaluation of total elastin, tropoelastin, collagen types I, III, and VII, and newly synthesized collagen. Noticeable clinical and histological improvement was observed after electro-optical synergy treatment. A statistically significant increase in the means of collagen types I, III, and VII, as well as newly synthesized collagen, together with increased levels of tropoelastin, were detected, while the mean level of total elastin was significantly decreased at the end of treatment and three months post-treatment. Electro-optical synergy is an effective treatment for contouring facial skin laxity. This modality stimulates the repair processes and reverses the clinical, as well as the histopathological, signs of aging with the advantage of being a relatively risk-free procedure with minimal patient recovery time.
Sigma-Aldrich
Elastic Stain