- [Constipation syndrome: methods for measuring the speed of intestinal transit].
[Constipation syndrome: methods for measuring the speed of intestinal transit].
Revista de gastroenterologia del Peru : organo oficial de la Sociedad de Gastroenterologia del Peru (2012-04-06)
Raúl León-Barúa
PMID22476180
ABSTRACT
In the present article, I remind what I presented, in a symposium performed in our Society on the constipation syndrome, in relation with the definition and the determinant factors of this syndrome, and, in addition, with the methods we have created to determine physiologically the velocity of the intestinal transit, specially colonic.