Merck
  • Different mechanisms are involved in apoptosis induced by melanoma gangliosides on human monocyte-derived dendritic cells.
Different mechanisms are involved in apoptosis induced by melanoma gangliosides on human monocyte-derived dendritic cells.
Glycobiology (2009-02-26)
Karim Bennaceur, Iuliana Popa, Jessica Alice Chapman, Camille Migdal, Josette Péguet-Navarro, Jean-Louis Touraine, Jacques Portoukalian
ABSTRACT
Tumor escape is linked to multiple mechanisms, notably the liberation, by tumor cells, of soluble factors that inhibit the function of dendritic cells (DC). We have shown that melanoma gangliosides impair DC differentiation and induce their apoptosis. The present study was aimed to give insight into the mechanisms involved. DC apoptosis was independent of the catabolism of gangliosides since lactosylceramide did not induce cell death. Apoptosis induced by GM3 and GD3 gangliosides was not blocked by inhibitors of de novo ceramide biosynthesis, whereas the acid sphingomyelinase inhibitor desipramine only prevented apoptosis induced by GM3. Furthermore, our results suggest that DC apoptosis was triggered via caspase activation, and it was ROS dependent with GD3 ganglioside, suggesting that GM3 and GD3 induced apoptosis through different mechanisms.
Sigma-Aldrich
L-Cycloserine