MilliporeSigma
  • Quantitative evaluation of chromosomal rearrangements in gene-edited human stem cells by CAST-Seq.
Quantitative evaluation of chromosomal rearrangements in gene-edited human stem cells by CAST-Seq.
Cell stem cell (2021-02-25)
Giandomenico Turchiano, Geoffroy Andrieux, Julia Klermund, Georges Blattner, Valentina Pennucci, Melina El Gaz, Gianni Monaco, Sushmita Poddar, Claudio Mussolino, Tatjana I Cornu, Melanie Boerries, Toni Cathomen
ABSTRACT
Genome editing has shown great promise for clinical translation but also revealed the risk of genotoxicity caused by off-target effects of programmable nucleases. Here we describe chromosomal aberrations analysis by single targeted linker-mediated PCR sequencing (CAST-Seq), a preclinical assay to identify and quantify chromosomal aberrations derived from on-target and off-target activities of CRISPR-Cas nucleases or transcriptional activator-like effector nucleases (TALENs), respectively, in human hematopoietic stem cells (HSCs). Depending on the employed designer nuclease, CAST-Seq detected translocations in 0%-0.5% of gene-edited human CD34+ HSCs, and up to 20% of on-target loci harbored gross rearrangements. Moreover, CAST-Seq detected distinct types of chromosomal aberrations, such as homology-mediated translocations, that are mediated by homologous recombination and not off-target activity. CAST-Seq is a sensitive assay able to identify and quantify unintended chromosomal rearrangements in addition to the more typical mutations at off-target sites. CAST-Seq analyses may be particularly relevant for therapeutic genome editing to enable thorough risk assessment before clinical application of gene-edited products.
Sigma-Aldrich
RAD51 Inhibitor B02, ≥98% (HPLC)