Merck
  • Acute effects of thyroid hormone analogs on sodium currents in neonatal rat myocytes.
Acute effects of thyroid hormone analogs on sodium currents in neonatal rat myocytes.
Journal of molecular and cellular cardiology (1999-05-18)
C J Huang, H M Geller, W L Green, W Craelius
ABSTRACT
We previously reported that T3(3,3',5-triiodo-L-thyronine) acutely increases sodium currents (INa) in neonatal rat myocytes. Here we compare the effects of several thyroid hormone analogs, including T4(3,3',5,5'-tetraiodo-L-thyronine), rT3(3,3',5'-triiodo-L-thyronine), D-T3(3,3',5-triiodo-D-thyronine), 3,5-T2(3,5-diiodo-L-thyronine), DIT (3,5-diiodo-L-tyrosine), MIT (3-monoiodo-L-tyrosine), tetrac (3,3',5,5'-tetraiodo-thyroacetic acid), triac (3, 3',5-triiodo-thyroacetic acid), and tyrosine, on INa in cultured neonatal rat myocytes (n ranged from 9 to 28 for each comparison). T4, T3, 3,5-T2, and DIT (10 n m) all increased current density relative to control to a similar degree: to 1.22+/-0.2, 1.21+/-0.03, 1.16+/-0.02 and 1.16+/-0.03, respectively, P<0.05. In contrast, thyroid hormone analogs with an altered side group of the inner iodophenyl ring, including tetrac, triac, and D-T3, had no effect on INa nor did rT3, MIT or tyrosine. Pretreatment with rT3 inhibited the effects of T4, T3, 3,5-T2, and DIT. Conversely, the dose-dependent inhibitory effect of amiodarone, an iodinated benzofuran derivative that antagonizes thyroid hormone actions, on INa was blocked when myocytes were pretreated with T3(100 n m, n=3), suggesting an interaction of T3 with amiodarone. The enhancement of INa by T3 and 3, 5-T2 could not be blocked by propranolol, suggesting that the effects are not mediated through beta -adrenergic signaling pathways. In conclusion, the present results suggest that the acute effects of thyroid hormone and analogs on cardiac INa are mediated by a non-genomic thyroid hormone receptor with a unique structure-activity relationship.
Sigma-Aldrich
3,5-Diiodo-L-thyronine, thyroid hormone analog