Merck
  • Placental Growth Factor Mediates Crosstalk Between Lung Cancer Cells and Tumor-Associated Macrophages in Controlling Cancer Vascularization and Growth.
Placental Growth Factor Mediates Crosstalk Between Lung Cancer Cells and Tumor-Associated Macrophages in Controlling Cancer Vascularization and Growth.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2018-07-11)
Changjun He, Kaibin Zhu, Xue Bai, Yingbin Li, Dawei Sun, Yaoguo Lang, Jinfeng Ning, Fenglin Sun, Changfa Qu, Shidong Xu
ABSTRACT
Assistance with tumor-associated vascularization is needed for the growth and invasion of non-small cell lung cancer (NSCLC). Recently, it was shown that placental growth factor (PLGF) expressed by NSCLC cells had a critical role in promoting the metastasis of NSCLC cells. However, the underlying molecular mechanisms remain elusive. Here, we first established a NSCLC model in mice that allows us not only to isolate tumor cells from non-tumor cells in the tumor, but also to trace tumor cells in living animals. Levels of PLGF, its unique receptor Flt-1, as well as transforming growth factor β1 (TGFβ1) was examined in tumor cells and tumor-associated macrophages (TAM) by RT-qPCR. A transwell well co-culture system and HUVEC assay were applied to study the crosstalk between NSCLC cells and TAM. NSCLC cells produced and secreted PLGF to signal to tumor-associated macrophages (TAM) through surface expression of Flt-1 on macrophages. In a transwell co-culture system, PLGF secreted by NSCLC cells triggered macrophage polarization to a TAM subtype that promote growth of NSCLC cells. Moreover, polarized TAM seemed to secrete TGFβ1 to enhance the growth of endothelial cells in a HUVEC assay. The cross-talk between TAM and NSCLC cells via PLGF/Flt-1 and TGFβ receptor signaling may promote the growth and vascularization of NSCLC.