Merck
  • Tiotropium Respimat Add-on Is Efficacious in Symptomatic Asthma, Independent of T2 Phenotype.
Tiotropium Respimat Add-on Is Efficacious in Symptomatic Asthma, Independent of T2 Phenotype.
The journal of allergy and clinical immunology. In practice (2017-11-28)
Thomas B Casale, Eric D Bateman, Mark Vandewalker, J Christian Virchow, Hendrik Schmidt, Michael Engel, Petra Moroni-Zentgraf, Huib A M Kerstjens
ABSTRACT
Adding tiotropium to existing inhaled corticosteroid (ICS) maintenance therapy with or without a long-acting β2-agonist (LABA) has been shown to be beneficial in patients with symptomatic asthma. To assess whether responses to tiotropium Respimat add-on therapy were influenced by patients' T2 status. In this exploratory study, data from 4 phase III trials were analyzed: once-daily tiotropium 5 μg or placebo as add-on to ICS + LABA (PrimoTinA-asthma; 2 replicate trials; NCT00772538/NCT00776984; n = 912); once-daily tiotropium 5 μg or 2.5 μg, twice-daily salmeterol 50 μg, or placebo as add-on to ICS (MezzoTinA-asthma; 2 replicate trials; NCT01172808/NCT01172821; n = 2100). The prespecified efficacy outcomes of these studies have been reported previously. Here, further exploratory subgroup analyses were performed to study whether these coprimary end points were influenced by serum IgE levels, blood eosinophil counts, and clinician judgment of allergic asthma. In addition, for the continuous parameters, namely, IgE and blood eosinophils, their influence on the treatment effect was modeled over the whole range of values. Tiotropium was efficacious in improving peak FEV1 within 3 hours postdose and trough FEV1, independent of T2 status. Tiotropium significantly reduced the risk of severe asthma exacerbations and asthma worsening, independent of T2 phenotype; Cox regression modeling supported a beneficial effect of tiotropium on exacerbations, independent of IgE levels or eosinophil counts. Numerical improvements in the 7-question Asthma Control Questionnaire (ACQ-7) responder rate with tiotropium versus placebo were observed in T2high and T2low patients; logistic regression modeling provided further evidence for improvement in ACQ-7 responder rates with tiotropium, independent of IgE levels or eosinophil counts. The results of our exploratory analyses suggest that the improvements seen with tiotropium Respimat as add-on to ICS ± LABA in patients with symptomatic asthma on lung function, exacerbation risk, and symptom control are independent of T2 phenotype.
Sigma-Aldrich
2-(Dimethylamino)ethanethiol hydrochloride, 95%