Ammonium Glycyrrhizate
Synonym(s):
Glycyrrhizic acid ammonium salt
Empirical Formula (Hill Notation):
C42H62O16 · NH3
CAS Number:
Molecular Weight:
839.96
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing