2-Butanone
Synonym(s):
Ethyl methyl ketone, MEK, Methyl ethyl ketone, 2-Butanone
Linear Formula:
C2H5COCH3
CAS Number:
Molecular Weight:
72.11
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Purity
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3