Dextrose
Synonym(s):
D-(+)-Glucose, Dextrose
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Analyte Suitability
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing