Skip to Content
Merck
Tropicamide impurity C European Pharmacopoeia (EP) Reference Standard

Ipratropium bromide Related Compound C

Tropicamide impurity C European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Tropic acid, 2-Phenylhydracrylic acid, 3-Hydroxy-2-phenylpropionic acid
Linear Formula:
C6H5CH(CH2OH)CO2H
CAS Number:
Molecular Weight:
166.17

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade