Skip to Content
Merck
Carboxymethylcellulose Sodium certified reference material, pharmaceutical secondary standard

Carboxymethylcellulose sodium

Carboxymethylcellulose Sodium certified reference material, pharmaceutical secondary standard
Synonym(s):
Carboxymethylcellulose sodium salt, Sodium carboxymethylcellulose
CAS Number:

Products

Brand
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade