SRT®-C SEC-2000 Gel Filtration Guard Column, 5 μm
Products
Product Category
Manufacturer
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing