Skip to Content
IE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Methyl-1-propanol
Synonym(s):
Isobutanol, Isobutyl alcohol
Linear Formula:
(CH
3
)
2
CHCH
2
OH
CAS Number:
78-83-1
Molecular Weight:
74.12
EC Number:
201-148-0
Products
Product Category
solvents (7)
Brand
Sigma-Aldrich (5)
Supelco (2)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Special Grade
ACS reagent (1)
Anhydrous (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
270466
2-Methyl-1-propanol,
suitable for HPLC, 99.5%
Expand
View Pricing
538132
2-Methyl-1-propanol,
99.5%
Expand
View Pricing
320048
2-Methyl-1-propanol,
ACS reagent, ≥99.0%
Expand
View Pricing
294829
2-Methyl-1-propanol,
anhydrous, 99.5%
Expand
View Pricing
1.00984
2-Methyl-1-propanol,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
1.00985
2-Methyl-1-propanol,
(isobutyl alcohol) EMPLURA
®
Expand
View Pricing
58448
2-Methyl-1-propanol,
BioUltra, for molecular biology, ≥99.5% (GC)
Expand
View Pricing