Thiazolyl Blue Tetrazolium Bromide
Synonym(s):
3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium bromide, MTT, Methylthiazolyldiphenyl-tetrazolium bromide
Empirical Formula (Hill Notation):
C18H16BrN5S
CAS Number:
Molecular Weight:
414.32
Products
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing