Cholesterol
Synonym(s):
3β-Hydroxy-5-cholestene, 5-Cholesten-3β-ol, Cholesterol, Cholesterol solution, Cholesterol, Highly Purified, Cholesterol from wool fat
Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Tissue
Quality Segment
Geographic Origin
Application
Technique
Showing 1-16 of 16
Compare
Product Number
Description
Pricing