Skip to Content
Merck
Potassium permanganate ACS reagent, ≥99.0%, low in mercury

Potassium permanganate

Potassium permanganate ACS reagent, ≥99.0%, low in mercury
Synonym(s):
Potassium permanganate
Empirical Formula (Hill Notation):
KMnO4
CAS Number:
Molecular Weight:
158.03

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Particle Size (µm)
Reagent Type
Special Grade