Skip to Content
Merck
All Photos(2)

Key Documents

SRP5095

Sigma-Aldrich

TTK, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ESK, FLJ38280, MPS1, MPS1L1, PYT

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

16-22 nmol/min·mg

mol wt

~130 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TTK(7272)

General description

TTK is a serine/threonine kinase that has been implicated in the regulation of centrosome duplication and mitotic checkpoint response. Overexpressing of dominant-negative TTK in human cell lines prevents centrosome duplication while active TTK accelerates centrosome reduplication in an osteosarcoma cell line. Disruption of TTK function leads to a combination of severe mitotic abnormalities and failure in centrosome duplication. TTK is required for stabilization and activation of p53 during spindle disruption. TTK phoshorylates the N-terminal domain of p53 at Thr18, and this phosphorylation disrupts the interaction with MDM2 and abrogates MDM2-mediated p53 ubiquitination.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Fu Huang et al.
Molecular and cellular biology, 29(11), 2935-2944 (2009-04-01)
Upon prolonged arrest in mitosis, cells undergo adaptation and exit mitosis without cell division. These tetraploid cells are either eliminated by apoptosis or arrested in the subsequent G(1) phase in a spindle checkpoint- and p53-dependent manner. p53 has long been
Harold A Fisk et al.
Proceedings of the National Academy of Sciences of the United States of America, 100(25), 14875-14880 (2003-12-06)
The mitotic spindle is essential for the maintenance of genetic stability, and in budding yeast its assembly and function depend on the Mps1 protein kinase. Mps1p is required for centrosome duplication and the spindle checkpoint. Several recent reports demonstrate that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service