Skip to Content
Merck
All Photos(1)

Key Documents

1.46185

Millipore

Lactose TTC A. w. Tergitol-7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

feature

ready-to-use

Quality Level

application(s)

food and beverages
industrial qc
microbiology
water monitoring

storage temp.

20-25°C

suitability

Escherichia coli

Application

Detection and enumeration of E. coli & coliform bacteria in water samples, Membrane filtration method

Packaging

90 mm plate (20 or 120 plates/box)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service