Merck
  • Design, synthesis and structure–activity relationship studies of morpholino-1H-phenalene derivatives that antagonize Mcl-1/Bcl-2.
Design, synthesis and structure–activity relationship studies of morpholino-1H-phenalene derivatives that antagonize Mcl-1/Bcl-2.
Bioorganic & medicinal chemistry (2015-01-15)
Xiangqian Li, Xiaomeng Liang, Ting Song, Pengchen Su, Zhichao Zhang
ABSTRACT
We report herein characteristic studies of Mcl-1 and Bcl-2 dual inhibitors. It was found that a protruding carbonyl group forming hydrogen bond with R263 plays a predominant role compared with the hydrophobic group that occupies the p2 pocket. A series of dual inhibitors representing different parts of the morpholino-1H-phenalene were designed, synthesized and evaluated.