Merck
  • Study of DDT and its derivatives DDD, DDE adsorption and degradation over Fe-SBA-15 at low temperature.
Study of DDT and its derivatives DDD, DDE adsorption and degradation over Fe-SBA-15 at low temperature.
Journal of environmental sciences (China) (2012-06-05)
Hailin Wang, Hua Tian, Zhengping Hao
ABSTRACT
Mesoporous SBA-15 with different Fe2O3 loading were synthesized by an in-situ coating progress for removals of dichlorodiphenyltrichloroethane (DDT) and its derivatives, i.e., 1,1-dichloro-2,2-bis-(p-chlorophenyl)ethane (DDD) and 1,1-dichloro-2,2-bis-(4-chloro -phenyl) ethane (DDE). The results from XRD (X-ray diffractometer), TEM (transmission electron microscopy) indicated that the iron could be well dispersed on SBA-15 within 6 wt.% Fe2O3 loading. Nitrogen adsorption-desorption tests indicated that the synthesized materials were characterized by ordered meso-structure, high surface area and large pore volume. DDTs were removed from aqueous media in 12-hr treatment and high removal efficiency of DDTs was achieved at over 94%. DDTs could be completely degraded at 350 degrees C under the existence of SBA-15 with 4 wt.% Fe2O3 loading. The final degradation products of DDT were dichlorobenzophenone (DCB) and bis-(4-chloro-phenyl) methane (DDM), suggesting a complete dechlorination from trichloromethyl.