Merck
Molecular mechanism of inhibition of nonclassical FGF-1 export.
Biochemistry (2005-11-23)
Dakshinamurthy Rajalingam, Thallapuranam Krishnaswamy S Kumar, Raffaella Soldi, Irene Graziani, Igor Prudovsky, Chin Yu
ABSTRACT
Fibroblast growth factor (FGF-1) lacks a signal sequence and is exported by an unconventional release mechanism. The nonclassical export of FGF-1 has been shown to be inhibited by an anti-allergic and anti-inflammatory drug, amlexanox (AMX). We investigate the molecular mechanism(s) underlying the inhibitory action of AMX on the release of FGF-1, using a variety of biophysical techniques including multidimensional NMR spectroscopy. AMX binds to FGF-1 and enhances its conformational stability. AMX binds to locations close to Cys30 and sterically blocks Cu(2+)-induced oxidation, leading to the formation of the homodimer of FGF-1. AMX-induced inhibition of the formation of the FGF-1 homodimer is observed both under cell-free conditions and in living cells. Results of this study suggest a novel approach for the design of drugs against FGF-1-mediated disorders.
Sigma-Aldrich
Amlexanox, ≥98% (HPLC)