Merck
  • Application of chemically induced dynamic nuclear polarization to the nitration of N-acetyltyrosine and to some reactions of peroxynitrite.
Application of chemically induced dynamic nuclear polarization to the nitration of N-acetyltyrosine and to some reactions of peroxynitrite.
Nitric oxide : biology and chemistry (2000-10-06)
A R Butler, T J Rutherford, D M Short, J H Ridd
ABSTRACT
By the observation of chemically induced dynamic nuclear polarization in (15)N NMR spectroscopy it has been shown that nitration of N-acetyltyrosine, even under acidic conditions, is largely a radical process. In the alkaline reaction of tyrosine with peroxynitrite the main products are nitrite and nitrate, both produced by a radical pathway, and tyrosine nitration is a minor reaction. It is suggested that tyrosine catalyzes the production of NO(*)(2) and HO(*) from peroxynitrite.
Product Description
Supelco
N-Acetyl-L-tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
N-Acetyl-L-tyrosine