Merck
  • Septin Defects Favour Symmetric Inheritance of the Budding Yeast Deceptive Courtship Memory.
Septin Defects Favour Symmetric Inheritance of the Budding Yeast Deceptive Courtship Memory.
International journal of molecular sciences (2023-02-12)
Fozia Akhtar, Bastien Brignola, Fabrice Caudron
ABSTRACT
Mnemons are prion-like elements that encode cellular memories of past cellular adaptations and do not spread to progenies during cell divisions. During the deceptive courtship in budding yeast, the Whi3 mnemon (Whi3mnem) condenses into a super-assembly to encode a mating pheromone refractory state established in the mother cell. Whi3mnem is confined to the mother cell such that their daughter cells have the ability to respond to the mating pheromone. Confinement of Whi3mnem involves its association with the endoplasmic reticulum membranes and the compartmentalization of these membranes by the lateral membrane diffusion barrier at the bud neck, the limit between the mother cell and the bud. However, during the first cell division after the establishment of the pheromone refractory state, this adaptation is more likely to be inherited by the daughter cell than in subsequent cell divisions. Here, we show that the first cell division is associated with larger daughter cells and cytokinesis defects, traits that are not observed in subsequent cell divisions. The cytoskeletal septin protein shows aberrant localisation in these divisions and the septin-dependent endoplasmic reticulum membrane diffusion barrier is weakened. Overall, these data suggest that cytokinesis defects associated with prolonged cell division can alter the confinement and inheritance pattern of a cellular memory.
Sigma-Aldrich
α1-Mating Factor acetate salt, ≥93% (HPLC), synthetic